Product Details

SNP ID
rs2304123
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:56375839 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGGATGTAGAATAATGAGCTTGAAT[C/T]GTCACATTTGGTAATAATCTTTTAT
Phenotype
MIM: 615600
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF542P PubMed Links

Gene Details

Gene
ZNF542P
Gene Name
zinc finger protein 542, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF582
Gene Name
zinc finger protein 582
There are no transcripts associated with this gene.

Gene
ZSCAN5A
Gene Name
zinc finger and SCAN domain containing 5A
There are no transcripts associated with this gene.

View Full Product Details