Product Details

SNP ID
rs11669504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45516057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTACTGACACCTCCCACCCAGG[C/T]TAAGTCAGACGTTGCCTTTTCCAGG
Phenotype
MIM: 601703
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VASP PubMed Links

Gene Details

Gene
VASP
Gene Name
vasodilator-stimulated phosphoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003370.3 Intron NP_003361.1
XM_005259199.1 Intron XP_005259256.1
XM_005259200.1 Intron XP_005259257.1
XM_017027200.1 Intron XP_016882689.1

View Full Product Details