Product Details
- SNP ID
-
rs8071879
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:48140095 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTAGTCTATGCCACACTTGTTGG[A/G]AAAGTTTACTCACTCACTGCTCCAA
- Phenotype
-
MIM: 604969
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SKAP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142423270] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SKAP1
- Gene Name
- src kinase associated phosphoprotein 1
View Full Product Details