Product Details

SNP ID
rs4794453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48107564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGATATCCATTCGCCGTCCCTCA[C/T]CCGGCACCTGCGTGCACTCTCCGGC
Phenotype
MIM: 604511 MIM: 614906
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CBX1 PubMed Links

Gene Details

Gene
CBX1
Gene Name
chromobox 1
There are no transcripts associated with this gene.

Gene
SNX11
Gene Name
sorting nexin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013323.2 7 Intron NP_037455.2
NM_152244.1 7 UTR 5 NP_689450.1
XM_005257260.3 7 Intron XP_005257317.1
XM_005257261.3 7 Intron XP_005257318.1
XM_005257262.3 7 UTR 5 XP_005257319.1
XM_011524697.2 7 UTR 5 XP_011522999.1

View Full Product Details