Product Details
- SNP ID
-
rs8737
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:2714722 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCAAGCATGGCCTGGCACAGTCCCC[A/G]GCACCAGGGATGCCCAGGCAGGATG
- Phenotype
-
MIM: 607862
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DIRAS1
PubMed Links
Gene Details
- Gene
- DIRAS1
- Gene Name
- DIRAS family GTPase 1
View Full Product Details