Product Details

SNP ID
rs2041518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93984426 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGAGAAGAGTTGGTGGACAACTA[C/T]GTCAAACCTTTTAGTACTGTTACAA
Phenotype
MIM: 605456
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BET1 PubMed Links

Gene Details

Gene
BET1
Gene Name
Bet1 golgi vesicular membrane trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317739.1 Intron NP_001304668.1
NM_005868.5 Intron NP_005859.1

View Full Product Details