Product Details

SNP ID
rs10197881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:102475256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAACATCTTTTGTGAAATCATAT[A/G]GACACTGATTGTGTTCAAGGTGATC
Phenotype
MIM: 600531
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC9A4 PubMed Links

Gene Details

Gene
SLC9A4
Gene Name
solute carrier family 9 member A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011552.3 Intron NP_001011552.2
XM_011511158.1 Intron XP_011509460.1

View Full Product Details