Product Details

SNP ID
rs35498978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:56655106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTGTTTTAACTTTATTTTGAT[A/G]TAATTATAGGTTCATAGGAAGTTGC
Phenotype
MIM: 603049 MIM: 613473
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TXNL1 PubMed Links

Gene Details

Gene
TXNL1
Gene Name
thioredoxin like 1
There are no transcripts associated with this gene.

Gene
WDR7
Gene Name
WD repeat domain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015285.2 Intron NP_056100.2
NM_052834.2 Intron NP_443066.2
XM_006722431.2 Intron XP_006722494.1
XM_011525888.1 Intron XP_011524190.1
XM_017025682.1 Intron XP_016881171.1
XM_017025683.1 Intron XP_016881172.1

View Full Product Details