Product Details

SNP ID
rs34633868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9721945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGACACTCAAGAGGGACTTTTT[C/T]CCTCAGATGTCTCAACAGCTGCCCT
Phenotype
MIM: 607467
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLECL1 PubMed Links

Gene Details

Gene
CLECL1
Gene Name
C-type lectin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253750.1 Intron NP_001240679.1
NM_001267701.1 Intron NP_001254630.1
NM_172004.3 Intron NP_742001.1
XM_011520574.2 Intron XP_011518876.1
XM_017018885.1 Intron XP_016874374.1

View Full Product Details