Product Details

SNP ID
rs2287002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2038820 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGTGGAAACGGCTTCGGGCCAGGC[A/G]GGGCTCCTGCTGTCTCAGGCCTTGG
Phenotype
MIM: 605069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MKNK2 PubMed Links

Gene Details

Gene
MKNK2
Gene Name
MAP kinase interacting serine/threonine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017572.3 2436 Intron NP_060042.2
NM_199054.2 2436 UTR 3 NP_951009.1

View Full Product Details