Product Details

SNP ID
rs11849403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75524111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCCCCTCTAGCCTGTGGGAGA[C/T]GTGGAAGGGAACAGGGCATGAGTTG
Phenotype
MIM: 612476
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BATF PubMed Links

Gene Details

Gene
BATF
Gene Name
basic leucine zipper ATF-like transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006399.3 Intron NP_006390.1

View Full Product Details