Product Details

SNP ID
rs11670635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18160766 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACATGACCAGGCCACTGCATGTG[C/T]TGTGCCCTCTCCCAGTTCTCCCTCC
Phenotype
MIM: 612258 MIM: 603157
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAST3 PubMed Links

Gene Details

Gene
MAST3
Gene Name
microtubule associated serine/threonine kinase 3
There are no transcripts associated with this gene.

Gene
PIK3R2
Gene Name
phosphoinositide-3-kinase regulatory subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005027.3 Intron NP_005018.1

View Full Product Details