Product Details

SNP ID
rs851149
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:223223695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACTCTGGGGATGAGGCCACCCA[C/T]ACTCCCTCCTGCACTCGGGGTCCTC
Phenotype
MIM: 615827
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUSD4 PubMed Links

Gene Details

Gene
SUSD4
Gene Name
sushi domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037175.2 Intron NP_001032252.1
NM_017982.3 Intron NP_060452.3
XM_005273169.1 Intron XP_005273226.1
XM_005273172.1 Intron XP_005273229.1
XM_006711408.1 Intron XP_006711471.1
XM_011509685.1 Intron XP_011507987.1
XM_011509687.1 Intron XP_011507989.1
XM_017001583.1 Intron XP_016857072.1
XM_017001584.1 Intron XP_016857073.1
XM_017001585.1 Intron XP_016857074.1
XM_017001586.1 Intron XP_016857075.1
XM_017001587.1 Intron XP_016857076.1
XM_017001588.1 Intron XP_016857077.1
XM_017001589.1 Intron XP_016857078.1

View Full Product Details