Product Details
- SNP ID
-
rs851150
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
25
- Location
-
Chr.1:223223538 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCGGGGCCTAAGGCACTCAAGCC[A/G]CCACTCACAGCTTCGTCATAGGACG
- Phenotype
-
MIM: 615827
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SUSD4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76683056] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SUSD4
- Gene Name
- sushi domain containing 4
View Full Product Details