Product Details
- SNP ID
-
rs7260314
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:32348187 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGAGGGCACTTTAGGGGAGGGAC[C/T]CTTGAGCTTATGTCCCCTGCCTTCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF507
PubMed Links
Gene Details
- Gene
- ZNF507
- Gene Name
- zinc finger protein 507
View Full Product Details