Product Details

SNP ID
rs2216107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:105015595 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGAATACCTATTTGGAAAATAGT[C/G]TGTTTTTTTCCTTCAATTTCTGAGA
Phenotype
MIM: 608444
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KMT2E PubMed Links

Gene Details

Gene
KMT2E
Gene Name
lysine methyltransferase 2E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018682.3 Intron NP_061152.3
NM_182931.2 Intron NP_891847.1
XM_005250493.1 Intron XP_005250550.1
XM_011516400.1 Intron XP_011514702.1
XM_017012435.1 Intron XP_016867924.1
Gene
KMT2E-AS1
Gene Name
KMT2E antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details