Product Details

SNP ID
rs8072775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40993489 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATCATGAAGAACTTCCTTAATTT[C/T]CAGAAAGATACCACCCAAGACTGAA
Phenotype
MIM: 616679
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT40 PubMed Links

Gene Details

Gene
KRT40
Gene Name
keratin 40
There are no transcripts associated with this gene.

Gene
KRTAP3-2
Gene Name
keratin associated protein 3-2
There are no transcripts associated with this gene.

Gene
KRTAP3-3
Gene Name
keratin associated protein 3-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033185.2 645 UTR 3 NP_149441.1

View Full Product Details