Product Details

SNP ID
rs13071292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187127587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTTTAGTGCCTATTTTAATGCA[C/T]AGTGAGATAACTGGTTTCCCTTTCC
Phenotype
MIM: 607547
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL39L PubMed Links

Gene Details

Gene
RPL39L
Gene Name
ribosomal protein L39 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052969.1 Intron NP_443201.1

View Full Product Details