Product Details

SNP ID
rs7486176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110059573 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACACCTAAGGCATTAAGTCTATA[C/T]AGGACTTCTGGGTCAAAGGGTGTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf76 PubMed Links

Gene Details

Gene
C12orf76
Gene Name
chromosome 12 open reading frame 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207435.2 30 Intron NP_997318.1
XM_017019297.1 30 Silent Mutation CTA,CTG L10L XP_016874786.1

View Full Product Details