Product Details

SNP ID
rs2241221
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:14806457 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AACCTTCTGCAATTCCTTTTGGATT[C/T]TCATAATGGCTTGGATGATGAGGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf60 PubMed Links

Gene Details

Gene
C12orf60
Gene Name
chromosome 12 open reading frame 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175874.3 2962 Intron NP_787070.2
XM_005253322.4 2962 Intron XP_005253379.1
XM_011520568.2 2962 Intron XP_011518870.1
XM_011520569.2 2962 Intron XP_011518871.1
XM_017018871.1 2962 Intron XP_016874360.1
XM_017018872.1 2962 Intron XP_016874361.1
XM_017018873.1 2962 Intron XP_016874362.1
XM_017018874.1 2962 Intron XP_016874363.1
Gene
SMCO3
Gene Name
single-pass membrane protein with coiled-coil domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013698.2 2962 Missense Mutation AAA,AGA K75R NP_001013720.2
XM_017019312.1 2962 Missense Mutation AAA,AGA K75R XP_016874801.1
Gene
WBP11
Gene Name
WW domain binding protein 11
There are no transcripts associated with this gene.

View Full Product Details