Product Details

SNP ID
rs1041445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41143810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGCGTGTTCTTGCATCATTATG[C/T]GTTCCCCCAGAACAGAAACGAGGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00323 PubMed Links

Gene Details

Gene
LINC00323
Gene Name
long intergenic non-protein coding RNA 323
There are no transcripts associated with this gene.

View Full Product Details