Product Details

SNP ID
rs4767440
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:116715580 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAATGACTTTTGGATCCTCCACCT[A/G]ATTTGGGTGTTTATGCCGCCTCGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf49 PubMed Links

Gene Details

Gene
C12orf49
Gene Name
chromosome 12 open reading frame 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024738.2 3018 UTR 3 NP_079014.1
XM_005253937.3 3018 UTR 3 XP_005253994.1

View Full Product Details