Product Details

SNP ID
rs1692141
Assay Type
Validated
NCBI dbSNP Submissions
61
Location
Chr.1:147758109 on Build GRCh38
Set Membership
JSNP Validated
Context Sequence [VIC/FAM]
TCAGTTCAGAAGGGACACGTCTTTC[T/C]TCTCTGAGCCTCCTTTGTTCCCACC
Phenotype
MIM: 121013
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
GJA5 PubMed Links

Gene Details

Gene
GJA5
Gene Name
gap junction protein alpha 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005266.6 3313 UTR 3 NP_005257.2
NM_181703.3 3313 UTR 3 NP_859054.1
XM_005272951.4 3313 UTR 3 XP_005273008.1
XM_017001044.1 3313 UTR 3 XP_016856533.1

View Full Product Details