Product Details

SNP ID
rs11607133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:14967652 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAACCAGTCATTCATCTTCCCAT[A/C]CCATCATACAAGGGCAGTCACCTTC
Phenotype
MIM: 114130
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CALCA PubMed Links

Gene Details

Gene
CALCA
Gene Name
calcitonin related polypeptide alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033952.2 Intron NP_001029124.1
NM_001033953.2 Intron NP_001029125.1
NM_001741.2 Intron NP_001732.1
XM_017018283.1 Intron XP_016873772.1
XM_017018284.1 Intron XP_016873773.1

View Full Product Details