Product Details
- SNP ID
-
rs2804253
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
58
- Location
-
Chr.1:108573447 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCTTGAAAAACTACAGTACCTCAT[A/G]TAGCCTTGGCTGGGAAAGGGAATAG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM102B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78873732] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM102B
- Gene Name
- family with sequence similarity 102 member B
View Full Product Details