Product Details

SNP ID
rs13202159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37007250 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGCTGTATTGAGGGCACTCATGG[A/C]AGGCAATGTTTCCTGTAGGCTTCAG
Phenotype
MIM: 605091
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FGD2 PubMed Links

Gene Details

Gene
FGD2
Gene Name
FYVE, RhoGEF and PH domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173558.3 Intron NP_775829.2
XM_011514372.2 Intron XP_011512674.1
XM_011514373.2 Intron XP_011512675.1
XM_017010429.1 Intron XP_016865918.1

View Full Product Details