Product Details
- SNP ID
-
rs10741130
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:27186515 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGGAATACTGCTCAGCACCTGAC[C/T]GTATCTGGATTTAGTCTGTAGCACA
- Phenotype
-
MIM: 616618
MIM: 608221
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACBD5
PubMed Links
Gene Details
- Gene
- ACBD5
- Gene Name
- acyl-CoA binding domain containing 5
- Gene
- MASTL
- Gene Name
- microtubule associated serine/threonine kinase like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001172303.2 |
3461 |
Silent Mutation |
ACC,ACT |
T873T |
NP_001165774.1 |
NM_001172304.2 |
3461 |
Silent Mutation |
ACC,ACT |
T834T |
NP_001165775.1 |
NM_001320756.1 |
3461 |
Silent Mutation |
ACC,ACT |
T877T |
NP_001307685.1 |
NM_001320757.1 |
3461 |
Silent Mutation |
ACC,ACT |
T878T |
NP_001307686.1 |
NM_032844.4 |
3461 |
Silent Mutation |
ACC,ACT |
T872T |
NP_116233.2 |
XM_005252631.3 |
3461 |
Silent Mutation |
ACC,ACT |
T835T |
XP_005252688.1 |
XM_005252632.3 |
3461 |
Silent Mutation |
ACC,ACT |
T801T |
XP_005252689.1 |
XM_006717519.3 |
3461 |
Silent Mutation |
ACC,ACT |
T844T |
XP_006717582.1 |
XM_006717520.3 |
3461 |
Intron |
|
|
XP_006717583.1 |
XM_011519751.2 |
3461 |
Silent Mutation |
ACC,ACT |
T541T |
XP_011518053.1 |
XM_017016852.1 |
3461 |
Silent Mutation |
ACC,ACT |
T838T |
XP_016872341.1 |
XM_017016853.1 |
3461 |
Silent Mutation |
ACC,ACT |
T829T |
XP_016872342.1 |
XM_017016854.1 |
3461 |
Intron |
|
|
XP_016872343.1 |
XM_017016855.1 |
3461 |
Intron |
|
|
XP_016872344.1 |
XM_017016856.1 |
3461 |
Intron |
|
|
XP_016872345.1 |
View Full Product Details