Product Details

SNP ID
rs3088247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:78777422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTATCTAGTTCCAGAACTTTCTC[A/C]TCACCCCAAATGGAAGCCCCATACA
Phenotype
MIM: 611542
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARSB PubMed Links

Gene Details

Gene
ARSB
Gene Name
arylsulfatase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000046.3 5076 UTR 3 NP_000037.2
NM_198709.2 5076 Intron NP_942002.1
XM_011543390.1 5076 UTR 3 XP_011541692.1
XM_011543391.2 5076 Intron XP_011541693.1
XM_011543392.2 5076 Intron XP_011541694.1
XM_011543393.1 5076 Intron XP_011541695.1
XM_017009471.1 5076 Intron XP_016864960.1

View Full Product Details