Product Details

SNP ID
rs12514445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55222205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTCTCAACAAGCGCGTCTCCCA[A/G]CGCTCTCTGGTTCTGGTCCGCCACC
Phenotype
MIM: 607752 MIM: 614086
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNO PubMed Links

Gene Details

Gene
CCNO
Gene Name
cyclin O
There are no transcripts associated with this gene.

Gene
MCIDAS
Gene Name
multiciliate differentiation and DNA synthesis associated cell cycle protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190787.1 754 Silent Mutation CTG,TTG L193L NP_001177716.1
XM_017009439.1 754 Silent Mutation CTG,TTG L62L XP_016864928.1

View Full Product Details