Product Details

SNP ID
rs590367
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:2231505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGGCCACCCGGCTTCTCTCTC[T/C]CTGTGGTTGTGAGCGCCGGGGTCCT
Phenotype
MIM: 164780
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SKI PubMed Links

Gene Details

Gene
SKI
Gene Name
SKI proto-oncogene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003036.3 Intron NP_003027.1
XM_005244775.3 Intron XP_005244832.1
XM_005244776.4 Intron XP_005244833.1
XM_017002128.1 Intron XP_016857617.1

View Full Product Details