Product Details
- SNP ID
-
rs10917682
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
44
- Location
-
Chr.1:163142414 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACATAGTACATAAAGTATTGAATAC[A/G]AAATTCTAAAGATAAACACAATTTT
- Phenotype
-
MIM: 603276
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RGS5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115870423] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RGS5
- Gene Name
- regulator of G-protein signaling 5
View Full Product Details