Product Details
- SNP ID
-
rs7908409
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:16519614 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AATACTGCAGACCCATGCGTAGCCC[G/T]TGTAGCGGCGCCGACCGGCTCAGGG
- Phenotype
-
MIM: 615227
MIM: 604446
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C1QL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144284413] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1QL3
- Gene Name
- complement C1q like 3
- Gene
- PTER
- Gene Name
- phosphotriesterase related
There are no transcripts associated with this gene.
View Full Product Details