Product Details

SNP ID
rs3746348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63555834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGTTCCTGACCTCGGACCTGAG[C/T]CCGCACCTGCTCAAGCGCCACCACG
Phenotype
MIM: 611265
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FNDC11 PubMed Links

Gene Details

Gene
FNDC11
Gene Name
fibronectin type III domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319152.1 247 Silent Mutation AGC,AGT S57S NP_001306081.1
NM_001319153.1 247 Silent Mutation AGC,AGT S57S NP_001306082.1
NM_024059.3 247 Silent Mutation AGC,AGT S57S NP_076964.1
XM_011529041.2 247 Silent Mutation AGC,AGT S57S XP_011527343.1
XM_017028060.1 247 Silent Mutation AGC,AGT S62S XP_016883549.1
Gene
HELZ2
Gene Name
helicase with zinc finger 2
There are no transcripts associated with this gene.

Gene
SRMS
Gene Name
src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites
There are no transcripts associated with this gene.

View Full Product Details