Product Details

SNP ID
rs511088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44281784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAATCAATCTTGGGTTTGAATC[A/G]TAGCCTTGCCACTTGCTACTGACTT
Phenotype
MIM: 186975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCTE1 PubMed Links

Gene Details

Gene
TCTE1
Gene Name
t-complex-associated-testis-expressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182539.3 Intron NP_872345.2
XM_005248872.1 Intron XP_005248929.1
XM_005248873.2 Intron XP_005248930.1
XM_005248874.2 Intron XP_005248931.1
XM_006715009.3 Intron XP_006715072.1
XM_011514336.2 Intron XP_011512638.1
XM_011514338.2 Intron XP_011512640.1
XM_011514339.2 Intron XP_011512641.1
XM_011514340.1 Intron XP_011512642.1
Gene
TMEM151B
Gene Name
transmembrane protein 151B
There are no transcripts associated with this gene.

View Full Product Details