Product Details
- SNP ID
-
rs324149
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:44229258 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ACACCCATAAGAGGACACATGCAAA[C/T]GGACACAAACACAGACGCCCTGTGC
- Phenotype
-
MIM: 602193
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC29A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148126154,rs7750723] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC29A1
- Gene Name
- solute carrier family 29 member 1 (Augustine blood group)
View Full Product Details