Product Details
- SNP ID
-
rs1023207
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:31805962 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ACACGCTCACAACAACAAATACAGA[A/C]TTTCTACCACAAAATTCAGGTGAGA
- Phenotype
-
MIM: 610577
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGAP12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12264870] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGAP12
- Gene Name
- Rho GTPase activating protein 12
View Full Product Details