Product Details

SNP ID
rs2244647
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:102726547 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGCAGGCAGTCTGCAACAGTGGGAC[C/G]TGCTCAGGTCCTCTGTGGTGGGCTT
Phenotype
MIM: 615570
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SFXN2 PubMed Links

Gene Details

Gene
SFXN2
Gene Name
sideroflexin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178858.4 Intron NP_849189.1
XM_011539261.2 Intron XP_011537563.1
XM_011539262.2 Intron XP_011537564.1
XM_011539263.2 Intron XP_011537565.1
XM_011539264.2 Intron XP_011537566.1
XM_017015663.1 Intron XP_016871152.1

View Full Product Details