Product Details

SNP ID
rs1891385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:6219845 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGAAATTGAGACAGTGGGATTTG[A/C]AAAGATTTTAATACTAACCCAAAAT
Phenotype
MIM: 608678
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IL33 PubMed Links

Gene Details

Gene
IL33
Gene Name
interleukin 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199640.1 Intron NP_001186569.1
NM_001199641.1 Intron NP_001186570.1
NM_001314044.1 Intron NP_001300973.1
NM_001314045.1 Intron NP_001300974.1
NM_001314046.1 Intron NP_001300975.1
NM_001314047.1 Intron NP_001300976.1
NM_001314048.1 Intron NP_001300977.1
NM_033439.3 Intron NP_254274.1
XM_011518061.2 Intron XP_011516363.1
XM_017015285.1 Intron XP_016870774.1

View Full Product Details