Product Details

SNP ID
rs7013202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39746972 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCCTCATACAATGAGTTGACAG[A/G]AACTTCCCCACCACTGTTTTCTGAA
Phenotype
MIM: 601533
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM2 PubMed Links

Gene Details

Gene
ADAM2
Gene Name
ADAM metallopeptidase domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278113.1 Intron NP_001265042.1
NM_001278114.1 Intron NP_001265043.1
NM_001464.4 Intron NP_001455.3
XM_005273468.1 Intron XP_005273525.1
XM_011544479.1 Intron XP_011542781.1
XM_011544480.1 Intron XP_011542782.1

View Full Product Details