Product Details

SNP ID
rs4274625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:190350309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTAATTTACTTTACAAAAAGGC[A/G]ATATAAGTTGACTCTTAGTGGAACA
Phenotype
MIM: 147263
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INPP1 PubMed Links

Gene Details

Gene
INPP1
Gene Name
inositol polyphosphate-1-phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128928.1 Intron NP_001122400.1
NM_002194.3 Intron NP_002185.1
XM_005246532.1 Intron XP_005246589.1

View Full Product Details