Product Details

SNP ID
rs2303615
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:9149717 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAGCACGCCCTATCAGAATTGTCA[A/G]AACTTCATGTAAATAGACAAGAATT
Phenotype
MIM: 604874 MIM: 176420
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLRG1 PubMed Links

Gene Details

Gene
KLRG1
Gene Name
killer cell lectin like receptor G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005810.3 Intron NP_005801.3
XM_017018682.1 Intron XP_016874171.1
XM_017018683.1 Intron XP_016874172.1
XM_017018684.1 Intron XP_016874173.1
XM_017018685.1 Intron XP_016874174.1
Gene
PZP
Gene Name
PZP, alpha-2-macroglobulin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002864.2 Intron NP_002855.2
XM_011520804.2 Intron XP_011519106.1
XM_011520805.2 Intron XP_011519107.1
XM_011520806.1 Intron XP_011519108.1
XM_011520807.2 Intron XP_011519109.1
XM_017019763.1 Intron XP_016875252.1
XM_017019764.1 Intron XP_016875253.1
XM_017019765.1 Intron XP_016875254.1
XM_017019766.1 Intron XP_016875255.1
XM_017019767.1 Intron XP_016875256.1

View Full Product Details