Product Details

SNP ID
rs8017640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76765379 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAGAGGTTTGGAAAAATTTGAAC[C/T]TCCCCCTCTTCAAACCACAGATCTC
Phenotype
MIM: 609011
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VASH1 PubMed Links

Gene Details

Gene
VASH1
Gene Name
vasohibin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014909.4 Intron NP_055724.1
XM_017021087.1 Intron XP_016876576.1
XM_017021088.1 Intron XP_016876577.1
XM_017021089.1 Intron XP_016876578.1
XM_017021090.1 Intron XP_016876579.1
XM_017021091.1 Intron XP_016876580.1

View Full Product Details