Product Details

SNP ID
rs12626123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:20021148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGTGAGGGTGCGGGTGGGCAG[A/G]CAAGAGGAGATAAAGCAGTTCAGTC
Phenotype
MIM: 610833
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAA20 PubMed Links

Gene Details

Gene
NAA20
Gene Name
N(alpha)-acetyltransferase 20, NatB catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016100.4 Intron NP_057184.1
NM_181527.3 Intron NP_852668.1
NM_181528.3 Intron NP_852669.1

View Full Product Details