Product Details
- SNP ID
-
rs36075160
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:58315096 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCACAGGGCCCTGGAGTCCGGAGG[A/G]GGAGCCGACATCACTTACTGCAGAA
- Phenotype
-
MIM: 612966
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PPP4R1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76191598] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPP4R1L
- Gene Name
- protein phosphatase 4 regulatory subunit 1 like (pseudogene)
There are no transcripts associated with this gene.
- Gene
- RAB22A
- Gene Name
- RAB22A, member RAS oncogene family
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020673.2 |
|
Intron |
|
|
NP_065724.1 |
View Full Product Details