Product Details
- SNP ID
-
rs6069225
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:38212475 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAAAATTAAAAAAAAGAAAATTT[G/T]CTTGACAACATACTCTTTATTCCCT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1755
PubMed Links
Gene Details
- Gene
- KIAA1755
- Gene Name
- KIAA1755
View Full Product Details