Product Details

SNP ID
rs12004487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120954403 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAAAGATAGTAATTTTGGGATTC[C/T]CAAAGAACTCACAAGTAGCCAGTTC
Phenotype
MIM: 120900
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C5 PubMed Links

Gene Details

Gene
C5
Gene Name
complement component 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317163.1 Intron NP_001304092.1
NM_001317164.1 Intron NP_001304093.1
NM_001735.2 Intron NP_001726.2
XM_011518980.2 Intron XP_011517282.1
XM_017015102.1 Intron XP_016870591.1
XM_017015103.1 Intron XP_016870592.1
XM_017015104.1 Intron XP_016870593.1

View Full Product Details