Product Details
- SNP ID
-
rs11615694
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:30930848 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCTTTCTCCCTGGGATGCTTTTCC[A/G]ACTGGCCATGCGACTTCCACTCACA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSPAN11
PubMed Links
Gene Details
- Gene
- TSPAN11
- Gene Name
- tetraspanin 11
View Full Product Details