Product Details

SNP ID
rs35710229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:36521685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACCCTCCGCCCTCCCACCTGCG[A/C]CCGCGTTTTTGTGCGGTCCCCACGG
Phenotype
MIM: 600635
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NKX2-1 PubMed Links

Gene Details

Gene
NKX2-1
Gene Name
NK2 homeobox 1
There are no transcripts associated with this gene.

Gene
NKX2-1-AS1
Gene Name
NKX2-1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SFTA3
Gene Name
surfactant associated 3
There are no transcripts associated with this gene.

View Full Product Details