Product Details

SNP ID
rs12587846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:36476034 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACATGCATGTGAACACTAATAAT[A/T]TTCCCAATGGATAAATGGGCAAAGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SFTA3 PubMed Links

Gene Details

Gene
SFTA3
Gene Name
surfactant associated 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101341.1 Intron NP_001094811.1

View Full Product Details