Product Details

SNP ID
rs741067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150803367 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGAGAGGAGTTGCATTTCCTGT[A/G]CTAAGCCTGCTCCTGGCTCAATCTC
Phenotype
MIM: 610334 MIM: 610385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM176A PubMed Links

Gene Details

Gene
TMEM176A
Gene Name
transmembrane protein 176A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018487.2 Intron NP_060957.2
XM_011516376.2 Intron XP_011514678.1
XM_011516377.2 Intron XP_011514679.1
XM_011516378.2 Intron XP_011514680.1
XM_017012393.1 Intron XP_016867882.1
Gene
TMEM176B
Gene Name
transmembrane protein 176B
There are no transcripts associated with this gene.

View Full Product Details